missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNAPac™ RP Columns

Product Code. 18078201 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
21.2 mm
Unit Size:
Each
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
18078201 10 mm 21.2 mm Each
15501693 100 mm 3 mm Each
15511693 50 mm 3 mm Each
15521693 10 mm 3 mm Pack of 2
15531693 100 mm 2.1 mm Each
15541693 50 mm 2.1 mm Each
15551693 10 mm 2.1 mm Pack of 2
18088201 10 mm 21.2 mm Pack of 2
18008211 150 mm 21.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 18078201 Supplier Thermo Scientific™ Supplier No. 95000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Get high resolution LC and LC-MS separation of oligonucleotides, including double-stranded (ds) DNA/RNA fragments, using these DNAPac reversed phase columns.

Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.

DNAPac RP reversed phase (RP) column is based on spherical wide-pore 4 μm polymer resin specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is highly stable to extreme pH (0-14), and temperature (5-110°C) and fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of small to large nucleic acids with low carry-over. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments:

  • Designed for ion-pair reversed-phase (IP-RP) LC and LC/MS separations of both oligonucleotides and double-stranded nucleic acids
  • High resolution and high throughput
  • Excellent MS compatibility
  • Wide operating pH range (0-14) and high temperature stability (up to 110°C)
  • Biocompatible hardware to eliminate analyte absorption
TRUSTED_SUSTAINABILITY

Specifications

Column Format Semi-preparative Guard Cartridge Holder
Diameter (Metric) 21.2 mm
Length (Metric) 10 mm
Quantity Each
Temperature up to 110°C
Product Line DNAPac
Type Preparative Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.