missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNASE Agar

Product Code. 11983462
Change view
Click to view available options
Description:
DNASE Agar
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11983462 DNASE Agar Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11983462 Supplier Thermo Scientific™ Supplier No. PO0128A
  • ONLINE EXCLUSIVE PRICE
    21.12 €
    16.53 € / Pack of 10
    Save 4.59 €
    21% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect the microbial deoxyribonuclease enzymes, particularly from staphylococci with this differential, ready-to-use medium.

Detect the microbial deoxyribonuclease enzymes, particularly from staphylococci with Thermo Scientific™ Oxoid™ DNASE Agar. A DNase (deoxyribonuclease) is any enzyme that catalyzes the cleavage of phosphodiester linkages of DNA leading to its degradation. The organisms producing DNase enzymes in sufficient quantities to hydrolyze DNA, will show clear zones around the colonies. The medium is mainly used in identification of staphylococci but may also be used for the detection of DNase activity in other organisms.

Use DNASE Agar for the diagnostic detection of deoxyribonuclease enzyme activity in staphylococci and other organisms.

  • Easily differentiates Staphylococcus aureus from Staphylococcus epidermidis and other Staphylococcus species due to the abundance of DNase production in Staphylococcus aureus. 1,2,3
  • Helps in the differentiation and identification of non-pigmented Serratia marcescens (positive reaction) from Klebsiella-Enterobacter (negative reaction) 4.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage  2° to 10°C
Description DNASE Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.