missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE1L3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92255-0.1ml
This item is not returnable.
View return policy
Description
DNASE1L3 Polyclonal antibody specifically detects DNASE1L3 in Mouse samples. It is validated for Western Blot
Specifications
| DNASE1L3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| deoxyribonuclease gamma, deoxyribonuclease I-like 3Liver and spleen DNase, deoxyribonuclease I-like III, DNAS1L3LSDDHP2, DNase gamma, DNase I homolog protein 2, DNase I homolog protein DHP2, DNase I-like 3, EC 3.1.21.-, EC 3.1.21.1, LS-DNase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-281 of human DNASE1L3 (NP_004935.1). MRICSFNVRSFGESKQEDKNAMDVIVKVIKRCDIILVMEIKDSNNRICPILMEKLNRNSRRGITYNYVISSRLGRNTYKEQYAFLYKEKLVSVKRSYHYHDYQDGDADVFSREPFVVWFQSPHTAVKDFVIIPLHTTPETSVKEIDELVEVYTDVKHRWKAENFIFMGDFNAGCSYVPKKAWKNIRLRTDPRFVWLIGDQEDTTVKKSTNCAYDRIVLRGQEIVSSVVPKSNSVFDFQKAYKLTEEEALDVSDHFPVEFKL | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 1776 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction