missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22652-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene shares considerable sequence similarity to, and is structurally related to DNase II. The latter is a well characterized endonuclease that catalyzes DNA hydrolysis in the absence of divalent cations at acidic pH. Unlike DNase II which is ubiquitously expressed, expression of this gene product is restricted to the salivary gland and lungs. The gene has been localized to chromosome 1p22.3 adjacent (and in opposite orientation) to the uricase pseudogene. Two transcript variants encoding different isoforms have been described for this gene.Specifications
| DNASE2B | |
| Polyclonal | |
| Unconjugated | |
| DNASE2B | |
| Deoxyribonuclease 2 beta, deoxyribonuclease II beta, DLAD, DNase II beta, DNase II like acid DNase, DNase2 like acid DNase, DNASE2B, Endonuclease DLAD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58511 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WZ79 | |
| DNASE2B | |
| DNASE2B Fusion Protein Ag18246 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction