missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1 Antibody (11H8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59694-25ug
This item is not returnable.
View return policy
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DNMT1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
| Mouse | |
| 180 kDa | |
| 25 μg | |
| Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators | |
| 1786 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 11H8 | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol with 0.05% Sodium Azide | |
| DNMT1 | |
| Raised against a synthetic peptide corresponding to amino acids 637-650 of human DNMT1 | |
| Protein G purified | |
| RUO | |
| Primary | |
| It will cross-react with mouse DNMT1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction