missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1 Antibody (4G11-C7), Novus Biologicals™
Mouse Monoclonal Antibody
178.00 € - 559.00 €
Specifications
| Antigen | DNMT1 |
|---|---|
| Clone | 4G11-C7 |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217211
|
Novus Biologicals
NBP2-59344 |
100 μg |
559.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666677
|
Novus Biologicals
NBP2-59344-25ug |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation.Specifications
| DNMT1 | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Fish, Zebrafish | |
| 1786 | |
| Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1 | |
| Primary | |
| Detects 180kDa. It will cross-react with mouse DNMT1. |
| 4G11-C7 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, ChIP Assay | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators | |
| AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
| DNMT1 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title