Learn More
Invitrogen™ TBK1 Recombinant Rabbit Monoclonal Antibody (24GB4650)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557763
 View more versions of this product
Product Code. 30275092
  • 198.00 € / 20µL
Description
TBK1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling. TBK1 is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-KB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-KB is associated with IKB proteins in the cell cytoplasm, which inhibit NF-KB activity. Phosphorylation of I-KB by IKB kinase (IKK) complex leads to degradation of I-KB and activation of NF-KB. The IKK complex contains IKKa, IKK beta, and IKKK. A novel IKK related kinase was recently identified and designated TBK1 (TANK-binding kinase 1), NAK (NF-KB-activating kinase), and T2K. NAK/TBK1 activates IKK beta through direct phosphorylation. NAK/TBK1 is activated by growth factors and PMA and mediates IKK and NF-kB activation in response to growth factors. NAK/TBK1 functions upstream of NIK and the IKK complex. NAK/TBK1 is also critical in protecting embryonic liver from apoptosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBK1
Recombinant Monoclonal
Unconjugated
TBK1
1200008B05Rik; AI462036; AW048562; FLJ11330; FTDALS4; NAK; NF-kappa-B-activating kinase; NF-kB-activating kinase; serine/threonine protein kinase TBK1; serine/threonine-protein kinase TBK1; T2K; TANK binding kinase 1; TANK-binding kinase 1; tbk; TBK1
Rabbit
Affinity Chromatography
RUO
29110, 299827, 56480
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4650
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UHD2, Q9WUN2
TBK1
A synthesized peptide derived from the N-term region of human NAK/TBK1 (aa 1-19).
20 μL
Primary
Human, Mouse, Rat