Learn More
Invitrogen™ DNMT3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA591951
Description
Positive Samples: Mouse spleen, Mouse skeletal muscle; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: LNVTLEHPLF VGGMCQNCKN CFLECAYQYD DDGYQSYCTI CCGGREVLMC GNNNCCRCFC VECVDLLVGP GAAQAAIKED PWNCYMCGHK GTYGLLRRRE DWPSRLQMFF ANNHDQEFDP PKVYPPVPAE KRKPIRVLSL FDGIATGLLV LKDLGIQVDR YIASEVCEDS ITVGMVRHQG KIMYVGDVRS VTQKHIQEWG P.
Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The Dnmt3 family members, Dnmt3a and Dnmt3b, are strongly expressed in ES cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of Dnmt3b gene occurs in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.
Specifications
| DNMT3A | |
| Polyclonal | |
| Unconjugated | |
| DNMT3A | |
| AU015806; DNA (cytosine-5-)-methyltransferase 3 alpha; DNA (cytosine-5)-methyltransferase 3A; DNA cytosine methyltransferase 3A2; DNA methyltransferase 3 alpha; DNA methyltransferase 3A; DNA methyltransferase HsaIIIA; DNA methyltransferase MmuIIIA; DNA MTase HsaIIIA; DNA MTase MmuIIIA; Dnmt3a; DNMT3A2; M.HsaIIIA; MmuIIIA; TBRS; W91664 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13435, 1788, 444984 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O88508, Q1LZ53, Q9Y6K1 | |
| DNMT3A | |
| A synthetic peptide corresponding to a sequence within amino acids 500-700 of human DNMT3A (NP_0720462). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.