missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DNMT3B Antibody (SR2574), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27699-50ul
This item is not returnable.
View return policy
Description
DNMT3B Monoclonal antibody specifically detects DNMT3B in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| DNMT3B | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR2574 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB | |
| A synthesized peptide derived from human DNMT3B (Uniprot #: Q9UBC3) | |
| 50 μL | |
| Cancer, Chromatin Research, DNA Repair, Epigenetics | |
| 1789 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction