missing translation for 'onlineSavingsMsg'
Learn More

DNQX, MedChemExpress

Product Code. 16491156
Encompass_Preferred
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16491156

Brand: Medchem Express HY15067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively).

DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively).

Specifications

Chemical Name or Material DNQX
Physical Form Powder
CAS 2379-57-9
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C8H4N4O6
For Use With (Application) Cancer-programmed cell death
Synonym FG 9041
Solubility Information DMSO : ≥ 35 mg/mL (138.81 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1C(NC2=CC([N+]([O-])=O)=C([N+]([O-])=O)C=C2N1)=O
Molecular Weight (g/mol) 252.14
Formula Weight 252.14
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.