missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNTTIP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
197.00 € - 463.00 €
Specifications
| Antigen | DNTTIP2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603870
|
Novus Biologicals
NBP2-92296-0.02ml |
0.02 mL |
197.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652391
|
Novus Biologicals
NBP2-92296-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNTTIP2 Polyclonal antibody specifically detects DNTTIP2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DNTTIP2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| acidic 82 kDa protein mRNA, deoxynucleotidyltransferase terminal-interacting protein 2, deoxynucleotidyltransferase, terminal, interacting protein 2, ERBP, estrogen receptor binding protein, FCF2, FLJ98849, HSU15552, LPTS-interacting protein 2, LPTS-RP2, MGC163494, TdIF2, tdT-interacting factor 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DNTTIP2 (NP_055412.2). MVVTRSARAKASIQAASAESSGQKSFAANGIQAHPESSTGSDARTTAESQTTGKQSLIPRTPKARKRKSRTTGSLPKGTEPSTDGETSEAESNYSVSEHHDTILRVTRRRQILIACSPVSSVRKKPKVTPTKESYTEEIVSEAESHVSGISRIVLPTEKTTGARRSKAKSLTDPSQESHTEAISDAETSSSDISFSGIATRRTRSMQRKLKAQTEKKDSKIVPGNEKQIVGTPVNSEDSDTRQTSHLQARSLSEINKPNFYNNDFDDDFSHRSSENILTVHEQANVESLKETKQNCKDLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 30836 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title