missing translation for 'onlineSavingsMsg'
Learn More

Dobutamine hydrochloride, MedChemExpress

Product Code. 16486906
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16486906 Supplier Medchem Express Supplier No. HY15746
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dobutamine hydrochloride is a synthetic catecholamine that acts on α1-AR, β1-AR, β2-AR (α-1, β-1 andβ-2 adrenoceptors). Dobutamine hydrochloride is a selective β1-AR agonist, relatively weak activity at α1-AR and β2-AR. Dobutamine hydrochloride can increase cardiac output and correct hypoperfusion.

Dobutamine hydrochloride is a synthetic catecholamine that acts on α1-AR, β1-AR, β2-AR (α-1, β-1 andβ-2 adrenoceptors). Dobutamine hydrochloride is a selective β1-AR agonist, relatively weak activity at α1-AR and β2-AR. Dobutamine hydrochloride can increase cardiac output and correct hypoperfusion.

Specifications

Chemical Name or Material Dobutamine hydrochloride
CAS 49745-95-1
Color White
Purity Grade Notes Research
Molecular Formula C18H24ClNO3
Quantity 100 mg
Solubility Information DMSO : ≥ 33 mg/mL (97.68 mM) ∣H2O : 20 mg/mL (59.20 mM; Need ultrasonic)
SMILES OC1=CC=C(CCNC(C)CCC2=CC=C(O)C=C2)C=C1O.[H]Cl
Molecular Weight (g/mol) 337.84
Formula Weight 337.84
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.