missing translation for 'onlineSavingsMsg'
Learn More

Docosapentaenoic acid 22n-3, MedChemExpress

Product Code. 16437487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437487 Supplier Medchem Express Supplier No. HY113159
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Docosapentaenoic acid (22n-3) is a component of phospholipids found in all animal cell membranes.

Docosapentaenoic acid (22n-3) is a component of phospholipids found in all animal cell membranes.

Specifications

Chemical Name or Material Docosapentaenoic acid 22n-3
Physical Form Liquid
CAS 24880-45-3
Color Colorless
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H34O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 50 mg/mL (151.29 mM; Need ultrasonic)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCCC(O)=O
Molecular Weight (g/mol) 330.5
Formula Weight 330.5
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.