missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl diphenyl phosphate, TRC

Product Code. 30180186
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180186 1 g 1g
30180218 100 mg 100mg
30180001 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180186 Supplier Toronto Research Chemicals Supplier No. TRCA5751331G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27460-02-2
Molecular Formula C24 H35 O4 P
Molecular Weight (g/mol) 418.506
Synonym Diphenyl lauryl phosphate, Dodecyl diphenyl phosphate
IUPAC Name dodecyl diphenyl phosphate
SMILES CCCCCCCCCCCCOP(=O)(Oc1ccccc1)Oc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H35O4P/c1-2-3-4-5-6-7-8-9-10-17-22-26-29(25,27-23-18-13-11-14-19-23)28-24-20-15-12-16-21-24/h11-16,18-21H,2-10,17,22H2,1H3
Formula Weight 418.227
Chemical Name or Material Dodecyl diphenyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.