missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG-1 / TMEM16A Mouse, Clone: DG1/1486, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 55107-MSM6-P1
This item is not returnable.
View return policy
Specifications
| DOG-1 / TMEM16A | |
| Cell Surface and Cytoplasm | |
| DG1/1486 | |
| Unconjugated | |
| NP_011543428 | |
| Mouse | |
| 114kDa | |
| Protein A/G Purified | |
| RUO | |
| 55107 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| TMEM16A | |
| Recombinant human DOG-1 protein fragment (aa 2-101) | |
| Gastrointestinal Stromal Tumor (GIST) or testicular germ cell tumor. Melanocytes in the basal layer of the epidermis and mast cells in the dermis of normal skin. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction