missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOHH Recombinant Rabbit Monoclonal Antibody (K01_1Q54)

Product Code. 30830448
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830448 50 μL
30824652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830448 Supplier Invitrogen™ Supplier No. MA564973
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: QYADGLEQLR GAPS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DOHH
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q54
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene DOHH
Gene Accession No. Q9BU89
Gene Alias Deoxyhypusine dioxygenase; deoxyhypusine hydroxylase; deoxyhypusine hydroxylase/monooxygenase; deoxyhypusine monooxygenase; DOHH; hDOHH; HEAT-like (PBS lyase) repeat containing 1; HEAT-like repeat-containing protein 1; HLRC1
Gene Symbols DOHH
Host Species Rabbit
Immunogen A synthetic peptide of human DOHH (aa 289-302).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83475
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.