missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOK3 Monoclonal Antibody (642316)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561487

Product Code. 30533839

  • 429.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DOK3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q7L591
DOK3
E. coli-derived recombinant human DOK3, Gly181-Lys271.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
642316
Unconjugated
DOK3
AI450713; Docking protein 3; Dok3; Dokl; DOK-L; Dok-like protein; downstream of tyrosine kinase 3; FLJ22570; FLJ39939; p62(dok)-like protein; p62Dok-like protein
Mouse
Protein A/G
RUO
79930
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.