missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dolichol Kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68873-25ul
This item is not returnable.
View return policy
Description
Dolichol Kinase Polyclonal antibody specifically detects Dolichol Kinase in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Dolichol Kinase | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| SEC59 Homolog, CDG1M, DK, DK1, Dolichol Kinase 1, EC 2.7.1.108, KIAA1094, SEC59, TMEM15, Transmembrane Protein 15 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YQNAKRSSSESKKHQAPTIARKY | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 22845 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction