missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir, MedChemExpress

Product Code. 16491066
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491066 200 mg 200mg
16481656 500 mg 500mg
16455578 10 mg 10mg
16451329 1 g 1g
16438277 5 mg 5mg
16436126 100 mg 100mg
16421747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16491066 Supplier Medchem Express Supplier No. HY13238
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dolutegravir (S/GSK1349572) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer. Dolutegravir (S/GSK1349572) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM).

Dolutegravir (S/GSK1349572) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer. Dolutegravir (S/GSK1349572) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM).

Specifications

Chemical Name or Material Dolutegravir
Physical Form Solid
CAS 1051375-16-6
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C20H19F2N3O5
For Use With (Application) COVID-19-anti-virus
Synonym S/GSK1349572
Solubility Information DMSO : 10 mg/mL (23.84 mM; Need ultrasonic and warming)
SMILES FC1=CC(F)=C(C=C1)CNC(C2=CN(C3=C(C2=O)O)C[C@@]([H])(N4C3=O)OCC[C@H]4C)=O
Molecular Weight (g/mol) 419.38
Formula Weight 419.38
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.