Learn More
Thermo Scientific™ Domed and Flat PCR Cap Strips
Product Code. 10249733
Change view
Click to view available options
Quantity:
120/Cs.
250/Cs.
Color:
Clear
Unit Size:
Pack of 120
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color Unit Size
10249733 250/Cs. Clear Pack of 250
10690174 120/Cs. Clear Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10249733 Supplier Thermo Scientific™ Supplier No. AB0784
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polypropylene
No. of Wells 8
Quantity 250/Cs.
Format Strips of 8 Flat Caps
Product Type PCR Plate Cap Strip
Certified RNase/DNase-free Yes
Plate Compatibility 96-well Plate