missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Flat PCR Cap Strips Promotion

Product Code. 15747486
Change view
Click to view available options
Quantity:
120/Cs.
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15747486 120/Cs. Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15747486 Supplier Thermo Scientific™ Supplier No. AB0866

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flat cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.

d

Highlights

  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
TRUSTED_SUSTAINABILITY

Specifications

Color Ultra Clear
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
Material Virgin Medical Grade Polypropylene
No. of Wells 96
No. per Case 120
No. per Pack 120
Quantity 120/Cs.
For Use With (Application) Facilitates sample labeling
Format Strips of 8 Flat Caps
Plate Compatibility 96-well PCR Plate, Standard and Low-profile PCR Tube Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.