missing translation for 'onlineSavingsMsg'
Learn More

Domiphen bromide, MedChemExpress

Product Code. 16461118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461118 Supplier Medchem Express Supplier No. HYB1467
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Domiphen bromide is a chemical antiseptic and a quaternary ammonium compound, used as a cationic surfactant.

Domiphen bromide is a chemical antiseptic and a quaternary ammonium compound, used as a cationic surfactant.

Specifications

Chemical Name or Material Domiphen bromide
CAS 538-71-6
Color White
Purity Grade Notes Research
Molecular Formula C22H40BrNO
Quantity 5 g
Solubility Information DMSO : 100 mg/mL (241.28 mM; Need ultrasonic) ∣H2O : 100 mg/mL (241.28 mM; Need ultrasonic)
SMILES CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]
Molecular Weight (g/mol) 414.46
Formula Weight 414.46
Percent Purity 99.49%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.