missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Dopa Decarboxylase/DDC Antibody Pair [HRP]

Product Code. p-7183091 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18280986

Brand: Novus Biologicals™ NBP27949515PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dopa Decarboxylase/DDC.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 109.38 pg/mL
Assay Sensitivity 156.25-10000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dopa Decarboxylase/DDC.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28
Gene ID (Entrez) 1644
Gene Symbol DDC
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Dopa Decarboxylase/DDC monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 2 mg/mL of rabbit anti-Human Dopa Decarboxylase/DDC monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 400 ng of recombinant Human Dopa Decarboxylase/DDC.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Neuroscience
    Storage Requirements Storage is content dependent.
    Target Dopa Decarboxylase/DDC
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.2 μg/mL in Dilution Buffer before use. 3. Standard: Reconstitute with 1 ml Dilution Buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 10000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.