missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D5R/DRD5 Antibody (SG4-D1b), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60019AF750
This item is not returnable.
View return policy
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Dopamine D5R/DRD5 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DRD5 | |
| Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-ProtNo. P25115] | |
| Primary | |
| Mouse, Rat, Human | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| SG4-D1b | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
| Mouse | |
| 0.1 mL | |
| 1816 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction