Learn More
Invitrogen™ Dopamine Transporter Recombinant Rabbit Monoclonal Antibody (24GB2160)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557262
 View more versions of this product
Product Code. 30274984
  • 146.00 € / 20µL
Description
Dopamine Transporter Recombinant Monoclonal Antibody for Western Blot, Flow
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dopamine Transporter
Recombinant Monoclonal
0.40 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q01959
Slc6a3
A synthesized peptide derived from human Dopamine Transporter (aa 150-215).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2160
Unconjugated
Slc6a3
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Rabbit
Affinity chromatography
RUO
6531
-20°C