missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doramapimod, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
Doramapimod (BIRB 796) is an orally active, highly potent p38 MAPK inhibitor, which has an IC50 for p38α=38 nM, for p38β=65 nM, for p38γ=200 nM, and for p38δ=520 nM. Doramapimod has picomolar affinity for p38 kinase (Kd=0.1 nM). Doramapimod also inhibits B-Raf with an IC50 of 83 nM.
Specifications
Specifications
| Chemical Name or Material | Doramapimod |
| CAS | 285983-48-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H37N5O3 |
| Quantity | 200 mg |
| Synonym | BIRB 796 |
| Solubility Information | DMSO : 125 mg/mL (236.89 mM; Need ultrasonic) ∣Ethanol : 33.33 mg/mL (63.17 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC(C(C)(C)C)=NN1C2=CC=C(C)C=C2)NC3=C4C=CC=CC4=C(OCCN5CCOCC5)C=C3 |
| Molecular Weight (g/mol) | 527.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction