missing translation for 'onlineSavingsMsg'
Learn More

Melford Doripenem

Product Code. 30018247
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
This item is not returnable. View return policy

Product Code. 30018247

Brand: Melford D628101.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 364622-82-2
  • Molecular Formula: C15H24N4O6S2 H2O
  • Molecular Weight: 438.52

Specifications

Chemical Name or Material Doripenem
CAS 364622-82-2
Quantity 1 g
Molecular Formula C15H24N4O6S2 - H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.