missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns

Product Code. 17457583 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
150 mm
500 mm
750 mm
Product Line:
Double nanoViper
Single nanoViper
Unit Size:
1 unit
Each
This item is not returnable. View return policy

Product Code. 17457583

Brand: Thermo Scientific™ DNV75500PN

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.

Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — Double nanoViper™ columns connect with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, Double nanoViper™
Diameter (Metric) 75 μm
Length (Metric) 500 mm
Max. Pressure 22,000 psi (1500 bar)
Particle Size 2 μm
pH 2 to 8
Pore Size 100 Å
Quantity Each
Surface Area 450 m2/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Double nanoViper
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Description Double nanoViper™ PepMap™ Neo 2 μm C18 75 μm X 500 mm
For Use With (Application) Peptide Mapping, LC-MS
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis