missing translation for 'onlineSavingsMsg'
Learn More

Doxepin Hydrochloride, MedChemExpress

Product Code. 16468588
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468588 10 g 10g
16431048 5 g 5g
16412269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468588 Supplier Medchem Express Supplier No. HYB0725
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Doxepin hydrochloride is an orally active tricyclic antidepressant agent.

Doxepin hydrochloride is a potent and selective histamine receptor H1 antagonist. Doxepin hydrochloride is also a potent CYP450 inhibitor and significantly inhibits CYP450 2C19 and 1A2. Doxepin inhibits reuptake of serotonin and norepinephrine as a tricyclic antidepressant.. Doxepin has therapeutic effects in atopic dermatitis,chronic urticarial,can improve cognitive processes, protect central nervous system.. Doxepin has also been proposed as a protective factor against oxidative stress..

Specifications

Chemical Name or Material Doxepin Hydrochloride
CAS 1229-29-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H22ClNO
Quantity 10 g
Solubility Information DMSO : ≥ 100 mg/mL (316.62 mM) ∣H2O : ≥ 50 mg/mL (158.31 mM)
SMILES CN(C)CC/C=C1C2=CC=CC=C2OCC3=CC=CC=C/13.Cl
Molecular Weight (g/mol) 315.84
Formula Weight 315.84
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.