missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride

Product Code. 15476139
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476139 10 mg 10mg
15486139 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476139 Supplier Thermo Scientific Alfa Aesar Supplier No. J64000.MA
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Doxorubicin hydrochloride, CAS # 25316-40-9, is the hydrochloride salt of doxorubicin, isolated from the bacterium Streptomyces peucetius, and is considered to be an anthracycline antibiotic with antineoplastic activity. | CAS: 25316-40-9 | C27H30ClNO11 | 579.98 g/mol

Antitumor antibiotic agent that inhibits DNA topoisomerase II.Doxorubicin hydrochloride is used as an antineoplastic, antibiotic agent that inhibits DNA topoisomerase. It also inhibits nucleic acid synthesis and induces apoptosis. It posses fluorescent property, which enable it for the measurement of drug efflux pump activities and the role of subcellular organelles (golgi and lysosome) in the sequestration of drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Doxorubicin hydrochloride, isolated from the bacterium Streptomyces peucetius, is an anthracycline antibiotic with antineoplastic activity and is related to daunorubicin
  • It can intercalate between base pairs in the DNA helix preventing DNA replication
  • It also inhibits the nucleic acid synthesis and can induce apoptosis
  • It can inhibit DNA topoisomerase II, resulting in an increased and stabilized cleavable enzyme-DNA linked complex during DNA replication, and prevents nucleotide strand ligation after double-strand breakage
  • Doxorubicin hydrochloride forms oxygen free radicals that result in cytotoxicity and lead to the lipid peroxidation of cell membrane lipids

Application

  • This compound displays a fluorescent property allowing the measurement of drug efflux pump activity and examining the role of subcellular organelles (Golgi complex and lysosome) in the sequestration of drugs
  • It has been used in cell viability assays
  • It is used as a drug in polymeric PLGA-based microparticulate drug delivery and to develop in vitro doxorubicin-resistant HepG2 cells (HepG2-DR) and K562 cells (K562-DR)
  • It is a substrate of the MRP1 transporter
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Melting Point 216°C (decomposition)
Color Orange-Red
Quantity 10 mg
Beilstein 4229251
Merck Index 14,3439
Solubility Information Soluble in water.
Formula Weight 579.99
Physical Form Solid
Chemical Name or Material Doxorubicin hydrochloride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Doxorubicin hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1A
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Doxorubicin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.