missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicinone, TRC

Product Code. 30004847
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004847 50 mg 50mg
17951455 5 mg 5mg
17910775 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004847 Supplier Toronto Research Chemicals Supplier No. TRCD55907050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24385-10-2
Molecular Formula C21 H18 O9
Molecular Weight (g/mol) 414.36
Synonym Doxorubicinone, (8S,10S)-6,8,10,11-Tetrahydroxy-8-(hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione, 5,12-Naphthacenedione, 7,8,9,10-tetrahydro-6,8,10,11-tetrahydroxy-8-(hydroxyacetyl)-1-methoxy-, (8S,10S)- (9CI), 5,12-Naphthacenedione, 7,8,9,10-tetrahydro-6,8,10,11-tetrahydroxy-8-(hydroxyacetyl)-1-methoxy-, (8S-cis)-, 5,12-Naphthacenedione, 8-glycoloyl-7,8,9,10-tetrahydro-6,8,10,11-tetrahydroxy-1-methoxy-, (8S,10S)- (8CI), (8S,10S)-7,8,9,10-Tetrahydro-6,8,10,11-tetrahydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,12-naphthacenedione, Adriamycin aglycone, Adriamycinone, Doxorubicin aglycone, Doxorubicinone, Epirubicin aglycone, Doxorubicin Hydrochloride Imp. D (EP), Epirubicine Hydrochloride Imp. A (EP)
IUPAC Name (7S,9S)-6,7,9,11-tetrahydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione
SMILES COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O)c4c(O)c3C(=O)c12)C(=O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H18O9/c1-30-11-4-2-3-8-14(11)20(28)16-15(17(8)25)18(26)9-5-21(29,12(24)7-22)6-10(23)13(9)19(16)27/h2-4,10,22-23,26-27,29H,5-7H2,1H3/t10-,21-/m0/s1
Formula Weight 414.0951
Percent Purity >95%
Chemical Name or Material Doxorubicinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.