missing translation for 'onlineSavingsMsg'
Learn More

Melford Doxycycline HCl

Product Code. 30018236
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018236 25 g 25g
30018234 10 g 10g
30018235 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018236 Supplier Melford Supplier No. D4302025.0
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Doxycycline HCl is a widely used antibiotic in the life science marketplace.

Doxycycline HCl is a widely used antibiotic in the life science marketplace.

It belongs to the tetracycline class of antibiotics and exhibits broad-spectrum activity against both gram-positive and gram-negative bacteria, as well as certain other microorganisms.

In the life science marketplace, Doxycycline HCl finds applications in various fields, including research and biotechnology. It is commonly used as an antibacterial agent in cell culture systems, where it helps prevent bacterial contamination in cell lines and maintain sterile conditions.

Doxycycline HCl is also utilized in molecular biology techniques such as gene expression studies and protein production. It can be added to media to select for cells containing specific genes or recombinant DNA constructs, as it inhibits protein synthesis in bacteria and other organisms. Its ability to induce gene expression or regulate protein production is exploited in certain research applications.

Specifications

Chemical Name or Material Doxycycline HCl
CAS 10592-13-9
Quantity 25 g
Molecular Formula C22H24N2O8-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.