missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hyclate, MP BIOMEDICALS

Product Code. 11486911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486911 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11486911 Supplier MP Biomedicals Supplier No. 0219895525
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Doxycycline is a relative of tetracycline reported to be more effective against sensitive organisms.

Doxycycline is a relative of tetracycline reported to be more effective against sensitive organisms.

Application Notes:

  • It is used to inhibit matrix metalloproteinases and has been used to inhibit the inflammatory response of Lyme Disease.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Antibiotic

Specifications

Physical Form Powder
CAS 24390-14-5
Color Yellow
Quantity 25 g
Molecular Weight (g/mol) 1025.89
Grade USP
Chemical Name or Material Doxycycline Hyclate
compliance-icons
Product Identifier
  • Doxycycline Hyclate, USP
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Doxycycline Hyclate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.