missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Doxycycline Hyclate ReadyMade™ Solution

Product Code. 30044033
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044033 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044033 Supplier TOKU-E™ Supplier No. D10910x1ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Doxycycline Hyclate ReadyMade™ Solution is provided as a sterile-filtered solution in DMSO.

  • CAS Number: 24390-14-5
  • Molecular Formula: C22H24N2O8 · HCl · 0.5 H2O · 0.5 C2H6O
  • Molecular Weight: 512.94 g/mol
  • Storage Conditions: -20°C, protect from light
  • Form: Liquid
  • Source: Streptomyces peucetius
  • Spectrum: Doxycycline Hyclate has broad-spectrum activity against Gram-positive and Gram-negative bacteria, and Mycoplasma (i.e. M. pneumoniae). Certain β-lactam resistant strains of vancomycin-resistant Enterococcus are also inhibited
  • Mechanism of Action: Tetracycline antimicrobials bind to the bacterial 30S ribosomal subunit interfering with tRNA/mRNA interaction, ultimately inhibiting protein synthesis. Tetracyclines can inhibit the matrix metalloprotease (MMP) enzyme family and inhibit mitochondrial biogenesis

Specifications

CAS 24390-14-5
Chemical Name or Material Doxycycline Hyclate
Color Yellow
Content And Storage -20°C, Protect from light
Description Doxycycline Hyclate ReadyMade™ Solution 100 mg/mL in 10% DMSO
Concentration 100 to 105 mg/mL
Form Clear, light yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in 10% DMSO
For Use With (Application) Microbiology, Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >95%
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces peucetius
Stem Cell Qualified Streptomyces peucetius
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.