missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hydrochloride, MP Biomedicals™

Product Code. 11420455 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420455 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420455 Supplier MP Biomedicals Supplier No. 0219504410
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

More active biologically than the corresponding 6b-epimer hydrochloride. Doxycycline Hydrochloride, MP Biomedicals is a broad spectrum antibiotic effective against gram-positive and gram-negative bacteria. It is a derivative of oxytetracycline used to inh

Doxycycline Hydrochloride is a synthetic oxytetracycline derivative and is a member of the tetracycline antibiotics group. It shows antiprotozoal properties and is a potent inhibitor of MMPs (matrix metallo-proteinases), such as type 1 collagenase. It is used to treat a variety of infections and for other biomedical applications.

  • Treats chronic prostatitis, sinusitis, pelvic inflammatory disease, acne, rosacea, and rickettsial infections
  • Inhibits the inflammatory response of Lyme disease
  • Used in prophylaxis of anthrax (caused by Bacillus anthracis)
  • Effective against Yersinia pestis
  • Inhibits collagen synthesis
  • Used in studies on wound healing and tissue remodeling
  • Eliminates Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs
  • Eliminates Ixodes scapularis ticks

Specifications

Physical Form Powder
CAS 10592-13-9
Concentration Working Concentration: It is typically used at a concentration of 20ug/mL and is stable in media at 37°C for approximately 5 days.
Quantity 10 g
Molecular Formula C22H25ClN2O8
MDL Number MFCD03427564
Synonym Biocamycin, Doxigalumicina, Doxycycline hyclate, Doxy-II, Midoxin, Novadox, Retens, Tetradox, Vibradox, Hydramycin, 2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride
Solubility Information Soluble in water (10mg/mL, clear, yellowish to yellowish-green solution) and DMSO (50mg/mL).
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
Molecular Weight (g/mol) 480.90
PubChem CID 54706018
Formula Weight 480.9
Grade Not Available
Show More Show Less
compliance-icons
Product Identifier
  • Doxycycline Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302+H312-Harmful if swallowed or in contact with skin
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.