missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™

Product Code. 10224633
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224633 1 g Amber Glass 1g
10619743 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224633 Supplier Fisher BioReagents Supplier No. BP26531
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Broad spectrum antibiotic

Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Type Doxycycline Hydrochloride
Physical Form Powder/Solid
Absorbance Inclusive between 300 at 349nm
Melting Point 210°C
Packaging Amber Glass
Potency Inclusive between 800μg/mg
Specific Rotation Inclusive between 105°
Water Inclusive between 1.4%
Color Yellow
Quantity 1 g
Assay Percent Range 95%
Formula Weight 480.9
Chemical Name or Material Doxycycline Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Doxycycline Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.