missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92787-0.02ml
This item is not returnable.
View return policy
Description
DP1 Polyclonal antibody specifically detects DP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human DP1 (NP_009042.1). MAKDAGLIEANGELKVFIDQNLSPGKGVVSLVAVHPSTVNPLGKQLLPKTFGQSNVNIAQQVVIGTPQRPAASNTLVVGSPHTPSTHFASQNQPSDSSPWSAGKRNRKGEKNGKGLRHFSMKVCEKVQRKGTTSYNEVADELVAEFSAADNHILPNESAYDQKNIRRRVYDALNVLMAMNIISKEKKEIKWIGLPTNSAQECQNLEVERQRRLERIKQKQSQLQELILQQIAFKNLVQRNRHAEQQASRPPPPNSVIHLPFIIVNTSKKTVIDCSISNDKFEYLFNFDNT | |
| 0.02 mL | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 7027 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction