missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP1 Antibody (SR1878), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22278-100ul
This item is not returnable.
View return policy
Description
DP1 Monoclonal antibody specifically detects DP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry
Specifications
| DP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1878 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
| A synthesized peptide derived from human DP1 (Uniprot #: Q14186) | |
| 100 μg | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 7027 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction