missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS (10X), calcium, magnesium
GREENER_CHOICE

Product Code. 11550466
Change view
Click to view available options
Quantity:
500mL
10 x 500 mL
Unit Size:
5000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11550466 500mL 500mL
11560466 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11550466 Supplier Gibco™ Supplier No. 14080048
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

DPBS (10X), calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility (14080-055). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 4.3 to 4.7
Physical Form Liquid
Quantity 500mL
Includes No Sodium Pyruvate
Osmolality 2700 to 3200 mOsm/kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.