missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility (14080-055). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility (14080-055). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 10 X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 4.3 to 4.7 |
| Physical Form | Liquid |
| Quantity | 10 x 500 mL |
| Includes | No Sodium Pyruvate |
| Osmolality | 2700 to 3200 mOsm/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
