missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, calcium, magnesium

Product Code. 11570456
Change view
Click to view available options
Quantity:
100mL
500mL
10 x 500 mL
20 x 100mL
6 x 1000 mL
Unit Size:
1 set
100mL
500mL
6 L
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11570456 100mL 100mL
11580456 500mL 500mL
11590456 20 x 100mL Pack of 20
11500466 10 x 500 mL 1 set
10572703 6 x 1000 mL 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11570456 Supplier Gibco™ Supplier No. 14040083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DPBS, calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 7 to 7.2
Physical Form Liquid
Quantity 100mL
Includes No Sodium Pyruvate
Osmolality 270 to 310 mOsm/kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.