missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DPBS, calcium, magnesium
DPBS, calcium, magnesium
Brand: Gibco™ 14040166
Additional Details : Weight : 0.10000kg
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
No Sodium Pyruvate | |
2° to 8°C | |
DPBS | |
Mammalian Cell Culture | |
7 to 7.2 | |
Liquid |
100 mL | |
1 X | |
20 x 100mL | |
270 to 310 mOsm/kg | |
For in-vitro diagnostic use |