missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13936
This item is not returnable.
View return policy
Description
DPP10 Polyclonal specifically detects DPP10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DPP10 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57628 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DPP10 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: GLLNRQCISCNFMKEQCTYFDASFSPMNQHFLLFCEGPRVPVVSLHSTDNPAKYFILESNSMLKEAILKKK | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction