missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPPI 1c hydrochloride

Description
Inhibitor of dipeptidyl peptidase IV (DPP-IV) (IC50 = 104 nM) that displays selectivity over enzymes with DPP-like activity (IC50 > 30 μM). Increases plasma GLP-1 levels and improves glucose tolerance in diabetic mice following oral glucose challenge.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 312.8 |
| Purity | >98% |
| Molecular Weight (g/mol) | 312.798 |
| Chemical Name or Material | DPPI 1c hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?