missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPII/QPP/DPP7 Rabbit anti-Mouse, Clone: 35, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90774-50UL
This item is not returnable.
View return policy
Description
DPPII/QPP/DPP7 Monoclonal specifically detects DPPII/QPP/DPP7 in Mouse samples. It is validated for ELISA.
Specifications
| DPPII/QPP/DPP7 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 35 | |
| ELISA 1:5000-1:10000 | |
| carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase | |
| Recombinant Mouse DPPII/QPP/DPP7 Protein | |
| 50 μg | |
| GPCR | |
| 29952 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction