missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPIV/CD26 Antibody (6G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26613-100ul
This item is not returnable.
View return policy
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| DPPIV/CD26 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 6G7 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| A synthesized peptide derived from Human DPPIV/CD26 [UniProt P27487] | |
| 100 μL | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| 1803 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction