missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPRX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560467
This item is not returnable.
View return policy
Description
Immunogen sequence: GVSTSVGLRN ADTLPRLPNA AHPIGLVYTG HRVPSFQLIL YPNLKVPAND FIGHRIVHFG CCRDPNIYCL YPILESQVCA PSFHSGSPAC SSNQSR Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 25%.
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the DPRX homeobox gene family. Evidence of mRNA expression has not yet been found for this gene. Multiple, related processed pseudogenes have been found which are thought to reflect expression of this gene in the germ line or embryonic cells.
Specifications
| DPRX | |
| Polyclonal | |
| Unconjugated | |
| DPRX | |
| Divergent paired-related homeobox; divergent-paired related homeobox; DPRX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 503834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NFQ7 | |
| DPRX | |
| Recombinant protein corresponding to Human DPRX. Recombinant protein control fragment (Product #RP-99208). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction