missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPYD Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 487.00 €
Specifications
| Antigen | DPYD |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635750
|
Novus Biologicals
NBP2-92308-0.02ml |
0.02 mL |
216.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671031
|
Novus Biologicals
NBP2-92308-0.1ml |
0.1 mL |
487.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPYD Polyclonal antibody specifically detects DPYD in Human, Mouse samples. It is validated for Western BlotSpecifications
| DPYD | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1806 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DHP, DHPDHase, dihydropyrimidine dehydrogenase, dihydropyrimidine dehydrogenase [NADP+], Dihydrothymine dehydrogenase, Dihydrouracil dehydrogenase, DPDMGC132008, EC 1.3.1.2, MGC70799 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human DPYD (NP_000101.2). MAPVLSKDSADIESILALNPRTQTHATLCSTSAKKLDKKHWKRNPDKNCFNCEKLENNFDDIKHTTLGERGALREAMRCLKCADAPCQKSCPTNLDIKSFITSIANKNYYGAAKMIFSDNPLGLTCGMVCPTSDLCVGGCNLYATEEGPINIGGLQQFATETLILAFSLMNHL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title