missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ X-plore™ 3500 Half-Mask Respirators

Product Code. 30243641
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30243641 Small Each
30243643 Medium Each
30243640 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30243641 Supplier Dräger™ Supplier No. R55351
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Made of soft and skin-friendly DrägerFlex material, X-plore™ 3500 features a drop-down strap design that enables the mask to be removed simply by releasing the fastener at the neck.

  • Modern, low-profile mask body design
  • Extended field of vision and room to easily use visors and shields
  • Can be worn comfortably under helmets and visors
  • Soft and skin-friendly DrägerFlex material
  • Soft, lightweight and flexible strap system
  • Flexi-Fit head harness conforms to the contours of head without pressure or hair entanglement
  • Innovative X-Guided cross-over strap system ensure even weight distribution and an even seal
  • 2-point bayonet connector with markings for insertion of the filter
  • Flexible nose seal fits securely with safety glasses and goggles
  • Drop-down strap design enables mask to be removed simply by releasing the fastener at the neck
  • Easy to disassemble, clean and reassemble

Specifications

Certifications/Compliance AS NZS 1716
Certifications (EN) EN 140
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
For Use With (Equipment) X-plore™ Bayonet Filters and Cartridges
Includes Drop-down harness system, Reasealable storage bag
Model R55351
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.