missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR6/TNFRSF21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
228.00 € - 315.00 €
Specifications
| Antigen | DR6/TNFRSF21 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642642
|
Novus Biologicals
NBP2-98249-100ul |
100 μg |
315.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693754
|
Novus Biologicals
NBP2-98249-50ul |
50 μg |
228.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DR6/TNFRSF21 Polyclonal antibody specifically detects DR6/TNFRSF21 in Mouse samples. It is validated for ELISASpecifications
| DR6/TNFRSF21 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| 0.2 um filtered solution in PBS | |
| 27242 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21 | |
| Produced in rabbits immunized with purified, recombinant Mouse DR6/TNFRSF21 (Accession#: NP_848704.1; Met 1-His 349) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title